Merck
  • Thermally reversible nanocellulose hydrogels synthesized via the furan/maleimide Diels-Alder click reaction in water.
Thermally reversible nanocellulose hydrogels synthesized via the furan/maleimide Diels-Alder click reaction in water.
International journal of biological macromolecules (2019-09-09)
Ricardo Klaus Kramer, Mohamed Naceur Belgacem, Antonio José Felix Carvalho, Alessandro Gandini
ABSTRACT
The study deals with the synthesis of thermally reversible hydrogels from modified cellulose nanofibers via the Diels-Alder "click" reaction in an aqueous medium. "Never-dried" cellulose fibres derived from hardwood were submitted to shearing and surface TEMPO-oxidation before being modified with furfurylamine. The ensuing pendant furan moieties were reacted with a water-soluble bismaleimide via Diels-Alder coupling at 65 °C to produce a hydrogel, whose deconstruction was induced by the corresponding retro-Diels-Alder reaction carried out at 95 °C. Differential scanning calorimetry and rheological measurement were used to characterize the hydrogels. These aqueous cellulosic materials should provide original applications in such areas as strong paper-based artefacts and biocompatible gels.
Sigma-Aldrich
Furfurylamine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
4,7,10-Trioxa-1,13-tridecanediamine, 97%
Pricing and availability is not currently available.