Merck
  • Structure-Activity Relationship Study on Col-003, a Protein-Protein Interaction Inhibitor between Collagen and Hsp47.
Structure-Activity Relationship Study on Col-003, a Protein-Protein Interaction Inhibitor between Collagen and Hsp47.
Chemical & pharmaceutical bulletin (2019-10-05)
Masahito Yoshida, Masazumi Saito, Shinya Ito, Koji Ogawa, Naoki Goshima, Kazuhiro Nagata, Takayuki Doi
ABSTRACT
This study demonstrates the structure-activity relationship of Col-003, a potent collagen-heat-shock protein 47 (Hsp47) interaction inhibitor. Col-003 analogues were successfully synthesized by Pd(0)-catalyzed cross-coupling reactions of 5-bromosalicylaldehyde derivatives with alkyl-metal species, and the inhibitory activities of the synthetic analogues were evaluated using surface plasmon resonance analysis (BIAcore). We succeeded in discovering two potent inhibitors that showed 85 and 81% inhibition at a concentration of 1.9 µM against the collagen-Hsp47 interaction. This indicates that elongation of an alkyl linker between two aromatic rings could considerably improve inhibitory activity due to the adjustment of a pendant phenyl moiety to an appropriate position, in addition to the hydrophobic interaction with an alkyl linker moiety.
Sigma-Aldrich
5-Bromosalicylaldehyde, 98%
Pricing and availability is not currently available.