Merck
  • A novel dual-function probe for recognition of Zn2+ and Al3+ and its application in real samples.
A novel dual-function probe for recognition of Zn2+ and Al3+ and its application in real samples.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2019-11-20)
Xue-Jiao Sun, Ting-Ting Liu, Na-Na Li, Shuang Zeng, Zhi-Yong Xing
ABSTRACT
A dual-function probe NAHH based on naphthalene was synthesized and characterized. Based on the combination effects derived from the inhabitation of photo-induced electron transfer (PET) and CN isomerization, probe NAHH achieved in the recognition of Zn2+ and Al3+ both through obvious fluorescence enhancement and color changes detected by naked eye, respectively. Probe NAHH showed high sensitivity with the limit of detection as low as 3.02 × 10-7 M for Zn2+ and 7.55 × 10-8 M for Al3+, indicated the capability of probe NAHH in trace detection for Zn2+ and Al3+. The binding ratio of NAHH with Zn2+ and Al3+ were all 1:1 determined by Job plot, and the corresponding association constant was calculated as 8.48 × 104 M-1 and 4.45 × 105 M-1, respectively. The mechanism was further confirmed by FT-IR, 1H NMR titration and ESI-MS analysis. Furthermore, probe NAHH was successfully applied in logic gate construction and the detection of Zn2+ and Al3+ in Songhua River and test stripe. Fluorescence imaging experiments confirmed that NAHH could be used to monitor Zn2+ in plant root.
Sigma-Aldrich
2-(2-Hydroxyphenyl)benzoxazole, 98%
Pricing and availability is not currently available.