Merck
  • Parallel assembly of actin and tropomyosin, but not myosin II, during de novo actin filament formation in live mice.
Parallel assembly of actin and tropomyosin, but not myosin II, during de novo actin filament formation in live mice.
Journal of cell science (2018-03-01)
Andrius Masedunskas, Mark A Appaduray, Christine A Lucas, María Lastra Cagigas, Marco Heydecker, Mira Holliday, Joyce C M Meiring, Jeff Hook, Anthony Kee, Melissa White, Paul Thomas, Yingfan Zhang, Robert S Adelstein, Tobias Meckel, Till Böcking, Roberto Weigert, Nicole S Bryce, Peter W Gunning, Edna C Hardeman
ABSTRACT
Many actin filaments in animal cells are co-polymers of actin and tropomyosin. In many cases, non-muscle myosin II associates with these co-polymers to establish a contractile network. However, the temporal relationship of these three proteins in the de novo assembly of actin filaments is not known. Intravital subcellular microscopy of secretory granule exocytosis allows the visualisation and quantification of the formation of an actin scaffold in real time, with the added advantage that it occurs in a living mammal under physiological conditions. We used this model system to investigate the de novo assembly of actin, tropomyosin Tpm3.1 (a short isoform of TPM3) and myosin IIA (the form of non-muscle myosin II with its heavy chain encoded by Myh9) on secretory granules in mouse salivary glands. Blocking actin polymerization with cytochalasin D revealed that Tpm3.1 assembly is dependent on actin assembly. We used time-lapse imaging to determine the timing of the appearance of the actin filament reporter LifeAct-RFP and of Tpm3.1-mNeonGreen on secretory granules in LifeAct-RFP transgenic, Tpm3.1-mNeonGreen and myosin IIA-GFP (GFP-tagged MYH9) knock-in mice. Our findings are consistent with the addition of tropomyosin to actin filaments shortly after the initiation of actin filament nucleation, followed by myosin IIA recruitment.
Sigma-Aldrich
Anti-Tropomyosin 4 Antibody, serum, Chemicon®
Pricing and availability is not currently available.