Merck
  • Clearance of solvents and small molecule impurities in antibody drug conjugates via ultrafiltration and diafiltration operation.
Clearance of solvents and small molecule impurities in antibody drug conjugates via ultrafiltration and diafiltration operation.
Biotechnology progress (2019-10-07)
Travis J Gates, Yaqi F Lyu, Xin Fang, Xiaoli Liao
ABSTRACT
Ultrafiltration and diafiltration (UF/DF) processes by tangential flow filtration (TFF) are frequently used for removal of solvents and small molecule impurities and for buffer exchange for biopharmaceutical products. Antibody-drug conjugates (ADCs) as an important class of biological therapeutics, carry unique solvents and small molecule impurities into the final UF/DF step as compared to standard antibody preparation. The production process of ADCs involves multiple chemical steps, for example, reduction and conjugation. The clearance of these solvents and small molecules by UF/DF, specifically the DF step, has been assessed and described herein. The rates of clearance for all the impurities in this study are close to the ideal clearance with no apparent interaction with either the protein or the TFF membrane and system. The effect of process variables during DF, such as pH, temperature, membrane loading, transmembrane pressure, and cross flow rate, has also been evaluated and found to have minimal impact on the clearance rate. These results demonstrate efficient clearance of solvents and small molecule impurities related to the ADC process by the DF process and provide a general data package to facilitate risk assessments based on the sieving factors and program specific needs.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Pricing and availability is not currently available.
Supelco
Ascentis® Express HILIC, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 3 mm
Pricing and availability is not currently available.