Merck
  • Preparation, Characterization, and Antimicrobial Activity of Cubosome Encapsulated Metal Nanocrystals.
Preparation, Characterization, and Antimicrobial Activity of Cubosome Encapsulated Metal Nanocrystals.
ACS applied materials & interfaces (2020-01-10)
Thomas G Meikle, Brendan P Dyett, Jamie B Strachan, Jacinta White, Calum J Drummond, Charlotte E Conn
ABSTRACT
Herein, we demonstrate a method for the functionalization of cubic phase lipid nanoparticles (cubosomes) with a series of magnetite (Fe3O4), copper oxide (Cu2O), and silver (Ag) nanocrystals, with prospective applications across a wide range of fields, including antimicrobial treatments. The resulting cubosomes are characterized using small-angle X-ray scattering and dynamic light scattering, demonstrating the retention of a typical cubic phase structure and particle size following nanocrystal encapsulation at concentrations up to 20% w/w. Cryogenic transmission electron microscopy reveals significant loading and association of each nanocrystal type with both monoolein- and phytantriol-based cubosomes. The antibiotic potential of these hybrid nanoparticles is demonstrated for the first time; cubosomes with embedded silver nanocrystals display a high level of antimicrobial activity against both Gram-positive and Gram-negative bacteria, with observed minimum inhibitory concentration values ranging from 15.6-250 μg/mL. Lastly, total internal reflection fluorescence microscopy is used to visualize cubosome-bacteria interactions, suggesting the involvement of particle interactions as a delivery mechanism.