Merck
  • Hyperglycemia does not affect tissue repair responses in shear stress-induced atherosclerotic plaques in ApoE-/- mice.
Hyperglycemia does not affect tissue repair responses in shear stress-induced atherosclerotic plaques in ApoE-/- mice.
Scientific reports (2018-05-16)
Sabrina Hsiung, Anki Knutsson, Jenifer Vallejo, Pontus Dunér, Suvi E Heinonen, Ann-Cathrine Jönsson-Rylander, Eva Bengtsson, Jan Nilsson, Anna Hultgårdh-Nilsson
ABSTRACT
The mechanisms responsible for macrovascular complications in diabetes remain to be fully understood. Recent studies have identified impaired vascular repair as a possible cause of plaque vulnerability in diabetes. This notion is supported by observations of a reduced content of fibrous proteins and smooth muscle cell mitogens in carotid endarterectomy from diabetic patients along with findings of decreased circulating levels of endothelial progenitor cells. In the present study we used a diabetic mouse model to characterize how hyperglycemia affects arterial repair responses. We induced atherosclerotic plaque formation in ApoE-deficient (ApoE-/-) and heterozygous glucokinase knockout ApoE-deficient mice (ApoE-/- GK+/-) mice with a shear stress-modifying cast. There were no differences in cholesterol or triglyceride levels between the ApoE-/- and ApoE-/- GK+/- mice. Hyperglycemia did not affect the size of the formed atherosclerotic plaques, and no effects were seen on activation of cell proliferation, smooth muscle cell content or on the expression and localization of collagen, elastin and several other extracellular matrix proteins. The present study demonstrates that hyperglycemia per se has no significant effects on tissue repair processes in injured mouse carotid arteries, suggesting that other mechanisms are involved in diabetic plaque vulnerability.
Sigma-Aldrich
Anti-Versican Antibody, a.a. 535-598 of mouse versican, Chemicon®, from rabbit
Pricing and availability is not currently available.