Merck
  • Selective Colorimetric "Turn-On" Probe for Efficient Engineering of Iminium Biocatalysis.
Selective Colorimetric "Turn-On" Probe for Efficient Engineering of Iminium Biocatalysis.
ACS omega (2020-02-18)
Lieuwe Biewenga, Michele Crotti, Mohammad Saifuddin, Gerrit J Poelarends
ABSTRACT
The efficient engineering of iminium biocatalysis has drawn considerable attention, with many applications in pharmaceutical synthesis. Here, we report a tailor-made iminium-activated colorimetric "turn-on" probe, specifically designed as a prescreening tool to facilitate engineering of iminium biocatalysis. Upon complexation of the probe with the catalytic Pro-1 residue of the model enzyme 4-oxalocrotonate tautomerase (4-OT), a brightly colored merocyanine-dye-type structure is formed. 4-OT mutants that formed this brightly colored species upon incubation with the probe proved to have a substantial activity for the iminium-based Michael-type addition of nitromethane to cinnamaldehyde, whereas mutants that showed no staining by the probe exhibited no or very low-level "Michaelase" activity. This system was exploited in a solid-phase prescreening assay termed as activated iminium colony staining (AICS) to enrich libraries for active mutants. AICS prescreening reduced the screening effort up to 20-fold. After two rounds of directed evolution, two artificial Michaelases were identified with up to 39-fold improvement in the activity for the addition of nitromethane to cinnamaldehyde, yielding the target γ-nitroaldehyde product with excellent isolated yield (up to 95%) and enantiopurity (up to >99% ee). The colorimetric activation of the turn-on probe could be extended to the class I aldolase 2-deoxy-d-ribose 5-phosphate aldolase, implicating a broader application of AICS in engineering iminium biocatalysis.
Sigma-Aldrich
all trans-Retinal, powder, ≥98%