Merck
  • In vitro Inhibitory Effects of Cynaroside on Human Liver Cytochrome P450 Enzymes.
In vitro Inhibitory Effects of Cynaroside on Human Liver Cytochrome P450 Enzymes.
Pharmacology (2019-10-07)
Lei Wang, Xiuju Ma, Jing Wang, Chang Li
ABSTRACT
Cynaroside is a biological component isolated from Lonicera japonica Thunb, and it possesses numerous pharmacological activities. However, whether cynaroside affects the activity of human liver cytochrome P450 (CYP) enzymes remains unclear. The purpose of this study was to investigate the effects of cynaroside on 8 major CYP isoforms in human liver microsomes (HLMs). In this study, the inhibitory effects of cynaroside on the 8 human liver CYP isoforms (i.e., 1A2, 3A4, 2A6, 2E1, 2D6, 2C9, 2C19, and 2C8) were investigated in vitro using HLMs. The results showed that cynaroside inhibited the activity of CYP1A2, 3A4, and 2C9, with IC50 values of 21.74, 15.88, and 16.58 μmol/L, respectively, but that other CYP isoforms were not affected. Enzyme kinetic studies showed that cynaroside was not only a noncompetitive inhibitor of CYP3A4 but also a competitive inhibitor of CYP1A2 and 2C9, with Ki values of 7.33, 11.60, and 8.09 μmol/L, respectively. In addition, cynaroside is a time-dependent inhibitor for CYP3A4 with Kinact/KI value of 0.049/11.62 μmol/L-1min-1. The in vitro studies of cynaroside with CYP isoforms indicate that cynaroside has the potential to cause pharmacokinetic drug interactions with other coadministered drugs metabolized by CYP1A2, 3A4, and 2C9. Further clinical studies are needed to evaluate the significance of this interaction.
Sigma-Aldrich
5-Carboxamidotryptamine maleate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Furafylline, ≥98% (HPLC)