Merck
  • Molecular profiling of thymoma with myasthenia gravis: Risk factors of developing myasthenia gravis in thymoma patients.
Molecular profiling of thymoma with myasthenia gravis: Risk factors of developing myasthenia gravis in thymoma patients.
Lung cancer (Amsterdam, Netherlands) (2019-12-07)
Ming-Ching Lee, Tzu-Hung Hsiao, Han-Ni Chuang, Li-Wen Lee, Pei-Ling Chi, Hui-Mei Tsai, Chien-Lin Mao, Chung-Ping Hsu
ABSTRACT
Thymoma is a rare epithelial tumor arising from the thymus in the anterior mediastinum. Nearly 50% of patients with thymoma develop myasthenia gravis, which is an indication of a poor long-term prognosis. Here, we identified specific and effective molecular markers for predicting in the development of myasthenia gravis patients with thymoma. We investigated molecular profiling based on RNA-sequencing (RNA-seq) for myasthenia gravis development in patients with thymoma. RNA was extracted from 34 patients with thymoma, 16 of whom had myasthenic and 18 of whom did not, and transcriptome profiles were analyzed through next-generation sequencing. We discovered 140 differential expressed genes associated with myasthenia gravis in thymoma patients. The four genes, hypoxia-inducible factor 3 alpha (HIF3A), insulin-like growth factor-binding protein 1, pyruvate dehydrogenase kinase, and Krüppel-like factor 15 were differentially expressed in patients with thymoma who has myasthenia gravis and were validated by quantitative polymerase chain reaction. HIF3A expression was significantly higher in patients with myasthenia gravis than in those without. HIF3A is aberrantly expressed in patient with thymoma who has myasthenia gravis and may be involved in the development of myasthenia gravis in thymoma patient.
Sigma-Aldrich
Anti-HIF3A antibody produced in rabbit, IgG fraction of antiserum
Pricing and availability is not currently available.