Merck
  • Epithelial tumor suppressor ELF3 is a lineage-specific amplified oncogene in lung adenocarcinoma.
Epithelial tumor suppressor ELF3 is a lineage-specific amplified oncogene in lung adenocarcinoma.
Nature communications (2019-11-30)
Katey S S Enfield, Erin A Marshall, Christine Anderson, Kevin W Ng, Sara Rahmati, Zhaolin Xu, Megan Fuller, Katy Milne, Daniel Lu, Rocky Shi, David A Rowbotham, Daiana D Becker-Santos, Fraser D Johnson, John C English, Calum E MacAulay, Stephen Lam, William W Lockwood, Raj Chari, Aly Karsan, Igor Jurisica, Wan L Lam
ABSTRACT
Gene function in cancer is often cell type-specific. The epithelial cell-specific transcription factor ELF3 is a documented tumor suppressor in many epithelial tumors yet displays oncogenic properties in others. Here, we show that ELF3 is an oncogene in the adenocarcinoma subtype of lung cancer (LUAD), providing genetic, functional, and clinical evidence of subtype specificity. We discover a region of focal amplification at chromosome 1q32.1 encompassing the ELF3 locus in LUAD which is absent in the squamous subtype. Gene dosage and promoter hypomethylation affect the locus in up to 80% of LUAD analyzed. ELF3 expression was required for tumor growth and a pan-cancer expression network analysis supports its subtype and tissue specificity. We further show that ELF3 displays strong prognostic value in LUAD but not LUSC. We conclude that, contrary to many other tumors of epithelial origin, ELF3 is an oncogene and putative therapeutic target in LUAD.
Sigma-Aldrich
Anti-ELF3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.