Merck
  • Chronic saponin treatment attenuates damage to the pancreas in chronic alcohol-treated diabetic rats.
Chronic saponin treatment attenuates damage to the pancreas in chronic alcohol-treated diabetic rats.
Journal of ginseng research (2017-10-13)
Mi Ran Choi, Su Min Kwak, Sol Hee Bang, Jo-Eun Jeong, Dai-Jin Kim
ABSTRACT
Chronic heavy alcohol consumption may raise the risk of developing type 2 diabetes mellitus. Saponins inhibit apoptosis of pancreatic islet cells and reduce lipid parameters. The present study was designed to investigate the effect of saponin on chronic ethanol-treated diabetic rats. Long-Evans Tokushima Fatty (LETO) and Otsuka Long-Evans Tokushima Fatty (OLETF) rats were pair-fed a Lieber-DeCarli diet with and without 5% ethanol for 12 wks. Two weeks after starting the pair-feeding with the Lieber-DeCarli diet, intraperitoneal injection of saponin was performed for 10 wks. To perform the experiments, rats were divided as follows: LETO-Control (LC), LETO-Ethanol (LE), LETO-Ethanol-Saponin (LES), OLETF-Control (OC), OLETF-Ethanol (OE), and OLETF-Ethanol-Saponin (OES). The weights of epididymal and mesenteric fat tissue in LES and OES rats were the lightest from among the LETO and OLETF groups, respectively. The secretion of alanine aminotransferase and cholesterol in OES rats decreased significantly compared to their secretion in OC and OE rats, respectively. The islets of the pancreas in LE and OE rats showed clean, unclear, and smaller morphology compared to those of LC, LES, OC, and OES rats. In addition, the expression of insulin in the islets of the pancreas in LC, LES, OC, and OES rats was higher than in LE and OE rats. Saponin may not only be helpful in alleviating the rapid progress of diabetes due to chronic alcohol consumption in diabetic patients, but may also show potential as an antidiabetic drug candidate for diabetic patients who chronically consume alcohol.
Sigma-Aldrich
Anti-Insulin Antibody, clone E11D7, clone E11D7, from mouse