Merck
  • Increased expression of kindlin 2 in luteinized granulosa cells correlates with androgen receptor level in patients with polycystic ovary syndrome having hyperandrogenemia.
Increased expression of kindlin 2 in luteinized granulosa cells correlates with androgen receptor level in patients with polycystic ovary syndrome having hyperandrogenemia.
Reproductive sciences (Thousand Oaks, Calif.) (2013-12-18)
Mei Yang, Juan Du, Danyu Lu, Caixia Ren, Huan Shen, Jie Qiao, Xi Chen, Hongquan Zhang
ABSTRACT
Hyperandrogenemia is the leading defect in patients with polycystic ovary syndrome (PCOS) and considered to be involved in the ovulation dysfunction of PCOS. During the process of ovulation, granulosa cells (GCs) undergo epithelial-mesenchymal transition (EMT), and integrin-interacting protein kindlin 2 is a well-known regulator in EMT. Therefore, our objective here was to compare the expression levels of kindlin 2 in luteinized GCs between patients with PCOS and control women and the relationship between kindlin 2 and PCOS pathogenesis. In this study, kindlin 2 expression was significantly increased in luteinized GCs from patients with PCOS, and kindlin 2 could be induced by testosterone both in vitro and in vivo. Meanwhile, kindlin 2 was positively correlated with androgen receptor (AR) in PCOS GCs. Taken together, kindlin 2 may play a role in luteinized GCs, especially in the case of excess androgen. Further studies are required to assess the specific role of kindlin 2 in follicular development and PCOS pathogenesis.
Sigma-Aldrich
Anti-Kindlin-2 Antibody, clone 3A3, clone 3A3, from mouse