Merck
  • Increased T-helper 17 cell differentiation mediated by exosome-mediated microRNA-451 redistribution in gastric cancer infiltrated T cells.
Increased T-helper 17 cell differentiation mediated by exosome-mediated microRNA-451 redistribution in gastric cancer infiltrated T cells.
Cancer science (2017-10-24)
Feng Liu, Zhouyan Bu, Feng Zhao, Daping Xiao
ABSTRACT
MicroRNA (miR)-451 is a cell metabolism-related miRNA that can mediate cell energy-consuming models by several targets. As miR-451 can promote mechanistic target of rapamycin (mTOR) activity, and increased mTOR activity is related to increased differentiation of T-helper 17 (Th17) cells, we sought to investigate whether miR-451 can redistribute from cancer cells to infiltrated T cells and enhance the distribution of Th17 cells through mTOR. Real-time PCR was used for detecting expression of miR-451 in gastric cancer, tumor infiltrated T cells and exosomes, and distribution of Th17 was evaluated by both flow cytometry and immunohistochemistry (IHC). Immunofluorescence staining was used in monitoring the exosome-enveloped miR-451 from cancer cells to T cells with different treatments, and signaling pathway change was analyzed by western blot. miR-451 decreased significantly in gastric cancer (GC) tissues but increased in infiltrated T cells and exosomes; tumor miR-451 was negatively related to infiltrated T cells and exosome miR-451. Exosome miR-451 can not only serve as an indicator for poor prognosis of post-operation GC patients but is also related to increased Th17 distribution in gastric cancer. miR-451 can redistribute from cancer cells to T cells with low glucose treatment. Decreased 5' AMP-activated protein kinase (AMPK) and increased mTOR activity was investigated in miR-451 redistributed T cells and the Th17 polarized differentiation of these T cells were also increased. Exosome miR-451 derived from tumor tissues can serve as an indicator for poor prognosis and redistribution of miR-451 from cancer cells to infiltrated T cells in low glucose treatment can enhance Th17 differentiation by enhancing mTOR activity.
Sigma-Aldrich
Anti-ROR gamma T Antibody, clone 6F3.1, clone 6F3.1, from mouse