Merck
  • Trophoblast-Specific Conditional Atg7 Knockout Mice Develop Gestational Hypertension.
Trophoblast-Specific Conditional Atg7 Knockout Mice Develop Gestational Hypertension.
The American journal of pathology (2018-08-31)
Aiko Aoki, Akitoshi Nakashima, Tae Kusabiraki, Yosuke Ono, Osamu Yoshino, Masanaga Muto, Keiichi Kumasawa, Tamotsu Yoshimori, Masahito Ikawa, Shigeru Saito
ABSTRACT
Hypertensive disorder of pregnancy (HDP) is a serious pregnancy complication that is life threatening to both the mother and fetus. Understanding HDP pathophysiology is important for developing medical treatments. This study demonstrates the involvement of autophagy deficiency in adverse maternal and fetal outcomes using trophoblast-specific autophagy related (Atg)7, an autophagy-related protein, knockout mice. Atg7 conditional knockout (cKO) placentas were significantly smaller than controls in the spongiotrophoblast layer but not the labyrinth layer, which significantly elevated blood pressure in dams. A marker of autophagy deficiency, sequestosome 1/p62, was accumulated in giant trophoblast cells and in the spongiotrophoblast layer, accompanying increased apoptosis. However, neither proteinuria in dams nor fetal growth restriction was observed. Regarding trophoblast function, the number of trophoblasts migrating into the maternal decidua was significantly reduced, and the wall/lumen ratio of the spiral arteries was significantly increased in cKO placentas, suggesting shallow trophoblast invasion and inadequate vascular remodeling. The relative expression of placental growth factor mRNA was significantly decreased in cKO placentas compared with the control, likely causing poor placentation; however, other factors were unchanged in cKO placentas. This is the first report of autophagy deficiency leading to impaired placentation complicated by maternal HDP attributable to trophoblast dysfunction, and it suggests that placental autophagy is required for normal placentation.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Pricing and availability is not currently available.
SAFC
NCTC 135 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.