Merck
  • Matrisome analysis of intrahepatic cholangiocarcinoma unveils a peculiar cancer-associated extracellular matrix structure.
Matrisome analysis of intrahepatic cholangiocarcinoma unveils a peculiar cancer-associated extracellular matrix structure.
Clinical proteomics (2019-11-07)
Guido Carpino, Diletta Overi, Fabio Melandro, Alessio Grimaldi, Vincenzo Cardinale, Sabina Di Matteo, Gianluca Mennini, Massimo Rossi, Domenico Alvaro, Vincenzo Barnaba, Eugenio Gaudio, Carmine Mancone
ABSTRACT
Intrahepatic cholangiocarcinoma (iCCA) is a malignancy that arises from the intrahepatic biliary tree, showing high mortality rates due to its late clinical presentation and limited treatment options. iCCA is characterized by a dense, reactive desmoplastic stroma marked by a dramatic accumulation of extracellular matrix (ECM). Although recent results strongly suggest a relationship between increasing desmoplastic stroma and the enhanced malignant behaviour of iCCA, the importance of ECM proteins in the pathogenesis of iCCA still have to be addressed. iCCA ECM fibrillar structural organization was characterized by histological analysis. ECM proteome profiles from decellularized iCCA and surrounding noncancerous tissues were analysed by nLC coupled to MALDI-TOF/TOF analysis. iCCA tissues displayed high levels of collagen fibers and low abundance of reticular and elastic fibers, suggesting stiffness and loss of polarity. The ECM proteome profiles of iCCA samples, when compared to those obtained from the surrounding noncancerous tissues showed a dismantling of the basement membrane, a reduced angiogenesis and a downregulation of oncosuppressive activity. In particular, we focused on the effects of the overexpression of collagen type III alpha 1 chain (COL3A1) in iCCA, thus providing evidences that COL3A1 promotes iCCA cells migration and is a component of tumor-associated aligned collagen. Overall, this study contributes to the understanding of molecular basis underlying desmoplasia in iCCA and indicates the type III collagen as a promising therapeutic target.
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
Human Collagen Type III
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Lubricin/Proteoglycan 4, clone 5C11 Antibody, clone 5C11, from mouse
Pricing and availability is not currently available.