Merck
  • Positive Feedback Loop of OCT4 and c-JUN Expedites Cancer Stemness in Liver Cancer.
Positive Feedback Loop of OCT4 and c-JUN Expedites Cancer Stemness in Liver Cancer.
Stem cells (Dayton, Ohio) (2016-06-25)
Kung-Kai Kuo, King-Teh Lee, Ker-Kong Chen, Ya-Han Yang, Ying-Chu Lin, Ming-Ho Tsai, Kenly Wuputra, Yen-Liang Lee, Chia-Chen Ku, Hiroyuki Miyoshi, Yukio Nakamura, Shigeo Saito, Chun-Chieh Wu, Chee-Yin Chai, Richard Eckner, Chen-Lung Steve Lin, Sophie S-W Wang, Deng-Chyang Wu, Chang-Shen Lin, Kazunari K Yokoyama
ABSTRACT
The network of stemness genes and oncogenes in human patient-specific reprogrammed cancer stem cells (CSCs) remains elusive, especially in liver cancer. HepG2-derived induced pluripotent stem cell-like cells (HepG2-iPS-like cells) were generated by introducing Yamanaka factors and the knockdown vector shTP53. They exhibited features of stemness and a higher tumorigenesis after xenograft transplantation compared with HepG2 cells. The cancerous mass of severe combined immunodeficiency (SCID) mice derived from one colony was dissected and cultured to establish reprogrammed HepG2-derived CSC-like cells (designated rG2-DC-1C). A single colony exhibited 42% occurrence of tumors with higher proliferation capacities. rG2-DC-1C showed continuous expression of the OCT4 stemness gene and of representative tumor markers, potentiated chemoresistance characteristics, and invasion activities. The sphere-colony formation ability and the invasion activity of rG2-DC-1C were also higher than those of HepG2 cells. Moreover, the expression of the OCT4 gene and the c-JUN oncogene, but not of c-MYC, was significantly elevated in rG2-DC-1C, whereas no c-JUN expression was observed in HepG2 cells. The positive-feedback regulation via OCT4-mediated transactivation of the c-JUN promoter and the c-JUN-mediated transactivation of the OCT4 promoter were crucial for promoting cancer development and maintaining cancer stemness in rG2-DC-1C. Increased expression of OCT4 and c-JUN was detected in the early stage of human liver cancer. Therefore, the positive feedback regulation of OCT4 and c-JUN, resulting in the continuous expression of oncogenes such as c-JUN, seems to play a critical role in the determination of the cell fate decision from iPS cells to CSCs in liver cancer. Stem Cells 2016;34:2613-2624.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, Alexa Fluor 488 Conjugate, clone TRA - 1-60, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit