Merck
  • s-Allyl cysteine, s-ethyl cysteine, and s-propyl cysteine alleviate β-amyloid, glycative, and oxidative injury in brain of mice treated by D-galactose.
s-Allyl cysteine, s-ethyl cysteine, and s-propyl cysteine alleviate β-amyloid, glycative, and oxidative injury in brain of mice treated by D-galactose.
Journal of agricultural and food chemistry (2011-05-10)
Shih-Jei Tsai, C Perry Chiu, Hui-Ting Yang, Mei-Chin Yin
ABSTRACT
The neuroprotective effects of s-allyl cysteine, s-ethyl cysteine, and s-propyl cysteine in D-galactose (DG)-treated mice were examined. DG treatment increased the formation of Aβ(1-40) and Aβ(1-42), enhanced mRNA expression of β-amyloid precursor protein (APP) and β-site APP cleavage enzyme 1 (BACE1), and reduced neprilysin expression in brain (P < 0.05); however, the intake of three test compounds significantly decreased the production of Aβ(1-40) and Aβ(1-42) and suppressed the expression of APP and BACE1 (P < 0.05). DG treatments declined brain protein kinase C (PKC) activity and mRNA expression (P < 0.05). Intake of test compounds significantly retained PKC activity, and the expression of PKC-α and PKC-γ (P < 0.05). DG treatments elevated brain activity and mRNA expression of aldose reductase (AR) and sorbitol dehydrogenase as well as increased brain levels of carboxymethyllysine (CML), pentosidine, sorbitol, and fructose (P < 0.05). Test compounds significantly lowered AR activity, AR expression, and CML and pentosidine levels (P < 0.05). DG treatments also significantly increased the formation of reactive oxygen species (ROS) and protein carbonyl and decreased the activity of glutathione peroxidase (GPX), superoxide dismutase (SOD), and catalase (P < 0.05); however, the intake of test compounds in DG-treated mice significantly decreased ROS and protein carbonyl levels and restored brain GPX, SOD, and catalase activities (P < 0.05). These findings support that these compounds via their anti-Aβ, antiglycative, and antioxidative effects were potent agents against the progression of neurodegenerative disorders such as Alzheimer's disease.
Sigma-Aldrich
L-Cysteine ethyl ester hydrochloride, 98%
Pricing and availability is not currently available.