Merck
  • Conjugated platinum(IV)-peptide complexes for targeting angiogenic tumor vasculature.
Conjugated platinum(IV)-peptide complexes for targeting angiogenic tumor vasculature.
Bioconjugate chemistry (2007-09-12)
Sumitra Mukhopadhyay, Carmen M Barnés, Ariel Haskel, Sarah M Short, Katie R Barnes, Stephen J Lippard
ABSTRACT
The integrins alpha vbeta3 and alpha vbeta5 and the membrane-spanning surface protein aminopeptidase N (APN) are highly expressed in tumor-induced angiogenesis, making them attractive targets for therapeutic intervention. Both integrins and APN recognize a broad range of peptides containing RGD (Arg-Gly-Asp) and NGR (Asn-Gly-Arg) motifs, respectively. Here, we describe the design, synthesis, and characterization of a series of mono- and difunctionalized platinum(IV) complexes in which a conjugated peptide motif, containing RGD, (CRGDC)c, (RGDfK)c, or NGR, is appended as a "tumor-homing device" to target tumor endothelial cells selectively over healthy cells. Platinum(IV)-peptide complexes with nonspecific amino acids or peptide moieties were prepared as controls. Concentration-response curves of these compounds were evaluated against primary proliferating endothelial cells and tumor cell lines and compared to those of cisplatin, a well-described platinum-based chemotherapeutic agent. The Pt(IV)-RGD conjugates were highly and specifically cytotoxic to cell lines containing alpha vbeta3 and alpha vbeta5, approaching the activity of cisplatin. The Pt(IV)-NGR complexes were less active than Pt(IV)-RGD-containing compounds but more active than nonspecific Pt-peptide controls. Integrin alpha vbeta3 mediated, at least in part, the anti-proliferative effect of a Pt(IV)-RGD conjugate, as demonstrated by a decreased inhibitory response when endothelial cells were either (1) incubated with an excess of alpha vbeta3/alpha vbeta5-specific RGD pentapeptides or (2) transfected with RNAi for beta 3, but not beta 1, integrins. These results suggest a rational approach to improved chemotherapy with Pt(IV)-peptide conjugates by selective drug delivery to the tumor compartment.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone B3B11, clone B3B11, Chemicon®, from mouse
Pricing and availability is not currently available.