Merck
  • Increased Cell Detachment Ratio of Mesenchymal-Type Lung Cancer Cells on pH-Responsive Chitosan through the β3 Integrin.
Increased Cell Detachment Ratio of Mesenchymal-Type Lung Cancer Cells on pH-Responsive Chitosan through the β3 Integrin.
Marine drugs (2019-11-28)
Chia-Hsiang Yen, Tai-Horng Young, Meng-Chi Hsieh, Li-Jen Liao, Tsung-Wei Huang
ABSTRACT
Chitosan is sensitive to environmental pH values due to its electric property. This study investigates whether the pH-responsive chitosan assay can provide a simple method to evaluate the aggressive behavior of cancer cells with cell detachment ratio. The epithelial-mesenchymal transition (EMT) is induced with transforming growth factor-β1 (TGF-β1) in the human non-small cell lung cancer cell line (A549). EMT-induced cells and untreated cells are cultured on chitosan substrates at pH 6.99 for 24 h, followed by pH 7.65 for 1 h. The cell detachment ratio (CDR) on pH-responsive chitosan rises with an increasing of the TGF-β1 concentration. The protein array reveals that the expression levels of the α2, α3, α5, β2, and β3 integrins are higher in EMT-induced A549 cells than in untreated cells. A further inhibition assay shows that adding β3 integrin blocking antibodies significantly decreases the CDR of EMT-induced cells from 32.7 ± 5.7% to 17.8 ± 2.1%. The CDR of mesenchymal-type lung cancer cells increases on pH-responsive chitosan through the β3 integrin. Notably, the CDR can be theoretically predicted according to the individual CDR on the pH-responsive chitosan surface, irrespective of heterogeneous cell mixture. The pH-responsive chitosan assay serves as a simple in vitro model to investigate the aggressive behavior of lung cancer including the heterogeneous cell population.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone 25E11, clone 25E11, Chemicon®, from mouse
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
α/β Integrin-mediated Cell Adhesion Array Combo Kit, colorimetic, The Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse