Merck
  • Dual Regulation of Gluconeogenesis by Insulin and Glucose in the Proximal Tubules of the Kidney.
Dual Regulation of Gluconeogenesis by Insulin and Glucose in the Proximal Tubules of the Kidney.
Diabetes (2017-06-21)
Motohiro Sasaki, Takayoshi Sasako, Naoto Kubota, Yoshitaka Sakurai, Iseki Takamoto, Tetsuya Kubota, Reiko Inagi, George Seki, Moritaka Goto, Kohjiro Ueki, Masaomi Nangaku, Takahito Jomori, Takashi Kadowaki
ABSTRACT
Growing attention has been focused on the roles of the proximal tubules (PTs) of the kidney in glucose metabolism, including the mechanism of regulation of gluconeogenesis. In this study, we found that PT-specific insulin receptor substrate 1/2 double-knockout mice, established by using the newly generated sodium-glucose cotransporter 2 (SGLT2)-Cre transgenic mice, exhibited impaired insulin signaling and upregulated gluconeogenic gene expression and renal gluconeogenesis, resulting in systemic insulin resistance. In contrast, in streptozotocin-treated mice, although insulin action was impaired in the PTs, the gluconeogenic gene expression was unexpectedly downregulated in the renal cortex, which was restored by administration of an SGLT1/2 inhibitor. In the HK-2 cells, the gluconeogenic gene expression was suppressed by insulin, accompanied by phosphorylation and inactivation of forkhead box transcription factor 1 (FoxO1). In contrast, glucose deacetylated peroxisome proliferator-activated receptor γ coactivator 1-α (PGC1α), a coactivator of FoxO1, via sirtuin 1, suppressing the gluconeogenic gene expression, which was reversed by inhibition of glucose reabsorption. These data suggest that both insulin signaling and glucose reabsorption suppress the gluconeogenic gene expression by inactivation of FoxO1 and PGC1α, respectively, providing insight into novel mechanisms underlying the regulation of gluconeogenesis in the PTs.
Sigma-Aldrich
Anti-IRS1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-IRS-2 Antibody, clone 9.5.2, clone 9.5.2, from mouse
Sigma-Aldrich
Anti-Sirt1 Antibody, clone 3H10.2, clone 3H10.2, Upstate®, from mouse