Merck
  • Functional Roles of Sex-Biased, Growth Hormone-Regulated MicroRNAs miR-1948 and miR-802 in Young Adult Mouse Liver.
Functional Roles of Sex-Biased, Growth Hormone-Regulated MicroRNAs miR-1948 and miR-802 in Young Adult Mouse Liver.
Endocrinology (2018-01-19)
Pengying Hao, David J Waxman
ABSTRACT
Sex-specific temporal patterns of pituitary growth hormone (GH) secretion determine the sex-biased transcription of hundreds of genes in the liver and impart important sex differences in liver physiology, metabolism, and disease. Sex differences in hepatic gene expression vary widely, ranging from less than twofold to >1000-fold in the mouse. Here, we use small RNA sequencing to discover 24 sex-biased mouse liver microRNAs (miRNAs), and then investigate the roles of two of these miRNAs in GH-regulated liver sex differences. Studies in prepubertal and young adult mice, and in mice in which pituitary hormones are ablated or where sex-specific hepatic GH signaling is dysregulated, demonstrated that the male-biased miR-1948 and the female-biased miR-802 are both regulated by sex-specific pituitary GH secretory patterns, acquire sex specificity at puberty, and are dependent on the GH-activated transcription factor STAT5 for their sex-specific expression. Both miRNAs are within genomic regions characterized by sex-biased chromatin accessibility. miR-1948, an uncharacterized miRNA, has essential features for correct Drosha/Dicer processing, generates a bona fide mature miRNA with strong strand bias for the 5p arm, and is bound by Argonaute in liver tissue, as is miR-802. In vivo studies using inhibitory locked nucleic acid sequences revealed that miR-1948-5p preferentially represses female-biased messenger RNAs (mRNAs) and induces male-biased mRNAs in male liver; conversely, miR-802-5p preferentially represses male-biased mRNAs and increases levels of female-biased mRNAs in female liver. Cytochrome P450 mRNAs were strongly enriched as targets of both miRNAs. Thus, miR-1948-5p and miR-802-5p are functional components of the GH regulatory network that shapes sex-differential gene expression in mouse liver.
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse
Pricing and availability is not currently available.