Merck
  • In vivo functional analysis of non-conserved human lncRNAs associated with cardiometabolic traits.
In vivo functional analysis of non-conserved human lncRNAs associated with cardiometabolic traits.
Nature communications (2020-01-04)
Xiangbo Ruan, Ping Li, Yi Chen, Yu Shi, Mehdi Pirooznia, Fayaz Seifuddin, Hiroshi Suemizu, Yasuyuki Ohnishi, Nao Yoneda, Megumi Nishiwaki, James Shepherdson, Abhilash Suresh, Komudi Singh, Yonghe Ma, Cheng-Fei Jiang, Haiming Cao
ABSTRACT
Unlike protein-coding genes, the majority of human long non-coding RNAs (lncRNAs) are considered non-conserved. Although lncRNAs have been shown to function in diverse pathophysiological processes in mice, it remains largely unknown whether human lncRNAs have such in vivo functions. Here, we describe an integrated pipeline to define the in vivo function of non-conserved human lncRNAs. We first identify lncRNAs with high function potential using multiple indicators derived from human genetic data related to cardiometabolic traits, then define lncRNA's function and specific target genes by integrating its correlated biological pathways in humans and co-regulated genes in a humanized mouse model. Finally, we demonstrate that the in vivo function of human-specific lncRNAs can be successfully examined in the humanized mouse model, and experimentally validate the predicted function of an obesity-associated lncRNA, LINC01018, in regulating the expression of genes in fatty acid oxidation in humanized livers through its interaction with RNA-binding protein HuR.
Sigma-Aldrich
RIPAb+ HuR - RIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
ChIPAb+ RNA Pol II - ChIP Validated Antibody and Primer Set, from mouse