Merck
  • Single-Cell RNA-Seq Uncovers a Robust Transcriptional Response to Morphine by Glia.
Single-Cell RNA-Seq Uncovers a Robust Transcriptional Response to Morphine by Glia.
Cell reports (2018-09-27)
Denis Avey, Sumithra Sankararaman, Aldrin K Y Yim, Ruteja Barve, Jeffrey Milbrandt, Robi D Mitra
ABSTRACT
Molecular and behavioral responses to opioids are thought to be primarily mediated by neurons, although there is accumulating evidence that other cell types play a prominent role in drug addiction. To investigate cell-type-specific opioid responses, we performed single-cell RNA sequencing (scRNA-seq) of the nucleus accumbens of mice following acute morphine treatment. Differential expression analysis uncovered unique morphine-dependent transcriptional responses by oligodendrocytes and astrocytes. We examined the expression of selected genes, including Cdkn1a and Sgk1, by FISH, confirming their induction by morphine in oligodendrocytes. Further analysis using RNA-seq of FACS-purified oligodendrocytes revealed a large cohort of morphine-regulated genes. The affected genes are enriched for roles in cellular pathways intimately linked to oligodendrocyte maturation and myelination, including the unfolded protein response. Altogether, our data illuminate the morphine-dependent transcriptional response by oligodendrocytes and offer mechanistic insights into myelination defects associated with opioid abuse.
Sigma-Aldrich
Milli-Mark® Anti-Galactocerebroside-FITC Antibody, clone mGalC, clone mGalC, Milli-Mark®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Pricing and availability is not currently available.