Merck
  • Impaired H3K36 methylation defines a subset of head and neck squamous cell carcinomas.
Impaired H3K36 methylation defines a subset of head and neck squamous cell carcinomas.
Nature genetics (2017-01-10)
Simon Papillon-Cavanagh, Chao Lu, Tenzin Gayden, Leonie G Mikael, Denise Bechet, Christina Karamboulas, Laurie Ailles, Jason Karamchandani, Dylan M Marchione, Benjamin A Garcia, Ilan Weinreb, David Goldstein, Peter W Lewis, Octavia Maria Dancu, Sandeep Dhaliwal, William Stecho, Christopher J Howlett, Joe S Mymryk, John W Barrett, Anthony C Nichols, C David Allis, Jacek Majewski, Nada Jabado
ABSTRACT
Human papillomavirus (HPV)-negative head and neck squamous cell carcinomas (HNSCCs) are deadly and common cancers. Recent genomic studies implicate multiple genetic pathways, including cell signaling, cell cycle and immune evasion, in their development. Here we analyze public data sets and uncover a previously unappreciated role of epigenome deregulation in the genesis of 13% of HPV-negative HNSCCs. Specifically, we identify novel recurrent mutations encoding p.Lys36Met (K36M) alterations in multiple H3 histone genes. histones. We further validate the presence of these alterations in multiple independent HNSCC data sets and show that, along with previously described NSD1 mutations, they correspond to a specific DNA methylation cluster. The K36M substitution and NSD1 defects converge on altering methylation of histone H3 at K36 (H3K36), subsequently blocking cellular differentiation and promoting oncogenesis. Our data further indicate limited redundancy for NSD family members in HPV-negative HNSCCs and suggest a potential role for impaired H3K36 methylation in their development. Further investigation of drugs targeting chromatin regulators is warranted in HPV-negative HNSCCs driven by aberrant H3K36 methylation.
Sigma-Aldrich
Anti-WHSC1/NSD2 Antibody, clone 29D1, clone 29D1, from mouse
Pricing and availability is not currently available.