Merck
  • Pharmacological inhibition of the spliceosome subunit SF3b triggers exon junction complex-independent nonsense-mediated decay.
Pharmacological inhibition of the spliceosome subunit SF3b triggers exon junction complex-independent nonsense-mediated decay.
Journal of cell science (2017-03-18)
Teresa Carvalho, Sandra Martins, José Rino, Sérgio Marinho, Maria Carmo-Fonseca
ABSTRACT
Spliceostatin A, meayamycin, and pladienolide B are small molecules that target the SF3b subunit of the spliceosomal U2 small nuclear ribonucleoprotein (snRNP). These compounds are attracting much attention as tools to manipulate splicing and for use as potential anti-cancer drugs. We investigated the effects of these inhibitors on mRNA transport and stability in human cells. Upon splicing inhibition, unspliced pre-mRNAs accumulated in the nucleus, particularly within enlarged nuclear speckles. However, a small fraction of the pre-mRNA molecules were exported to the cytoplasm. We identified the export adaptor ALYREF as being associated with intron-containing transcripts and show its requirement for the nucleo-cytoplasmic transport of unspliced pre-mRNA. In contrast, the exon junction complex (EJC) core protein eIF4AIII failed to form a stable complex with intron-containing transcripts. Despite the absence of EJC, unspliced transcripts in the cytoplasm were degraded by nonsense-mediated decay (NMD), suggesting that unspliced transcripts are degraded by an EJC-independent NMD pathway. Collectively, our results indicate that although blocking the function of SF3b elicits a massive accumulation of unspliced pre-mRNAs in the nucleus, intron-containing transcripts can still bind the ALYREF export factor and be transported to the cytoplasm, where they trigger an alternative NMD pathway.
Sigma-Aldrich
Anti-CWC22 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.