Merck
  • Bi-allelic ADPRHL2 Mutations Cause Neurodegeneration with Developmental Delay, Ataxia, and Axonal Neuropathy.
Bi-allelic ADPRHL2 Mutations Cause Neurodegeneration with Developmental Delay, Ataxia, and Axonal Neuropathy.
American journal of human genetics (2018-11-08)
Katharina Danhauser, Bader Alhaddad, Christine Makowski, Dorota Piekutowska-Abramczuk, Steffen Syrbe, Natalia Gomez-Ospina, Melanie A Manning, Anna Kostera-Pruszczyk, Claudia Krahn-Peper, Riccardo Berutti, Reka Kovács-Nagy, Mirjana Gusic, Elisabeth Graf, Lucia Laugwitz, Michaela Röblitz, Andreas Wroblewski, Hans Hartmann, Anibh M Das, Eva Bültmann, Fang Fang, Manting Xu, Ulrich A Schatz, Daniela Karall, Herta Zellner, Edda Haberlandt, René G Feichtinger, Johannes A Mayr, Thomas Meitinger, Holger Prokisch, Tim M Strom, Rafał Płoski, Georg F Hoffmann, Maciej Pronicki, Penelope E Bonnen, Susanne Morlot, Tobias B Haack
ABSTRACT
ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases, such as ADPRHL2, reverse the reaction. We used exome sequencing to identify five different bi-allelic pathogenic ADPRHL2 variants in 12 individuals from 8 families affected by a neurodegenerative disorder manifesting in childhood or adolescence with key clinical features including developmental delay or regression, seizures, ataxia, and axonal (sensori-)motor neuropathy. ADPRHL2 was virtually absent in available affected individuals' fibroblasts, and cell viability was reduced upon hydrogen peroxide exposure, although it was rescued by expression of wild-type ADPRHL2 mRNA as well as treatment with a PARP1 inhibitor. Our findings suggest impaired protein ribosylation as another pathway that, if disturbed, causes neurodegenerative diseases.
Sigma-Aldrich
Anti-pan-ADP-ribose binding reagent, from Escherichia coli
Sigma-Aldrich
Anti-ADPRHL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2