Merck
  • A Surveillance Mechanism Ensures Repair of DNA Lesions during Zygotic Reprogramming.
A Surveillance Mechanism Ensures Repair of DNA Lesions during Zygotic Reprogramming.
Cell (2016-12-06)
Sabrina Ladstätter, Kikuë Tachibana-Konwalski
ABSTRACT
Sexual reproduction culminates in a totipotent zygote with the potential to produce a whole organism. Sperm chromatin reorganization and epigenetic reprogramming that alter DNA and histone modifications generate a totipotent embryo. Active DNA demethylation of the paternal genome has been proposed to involve base excision and DNA repair-based mechanisms. The nature and consequence of DNA lesions generated during reprogramming are not known. Using mouse genetics and chemical biology, we discovered that Tet3-dependent zygotic reprogramming generates paternal DNA lesions that are monitored by a surveillance mechanism. In vivo structure-function rescue assays revealed that cohesin-dependent repair of paternal DNA lesions prevents activation of a Chk1-dependent checkpoint that delays mitotic entry. Culturing conditions affect checkpoint stringency, which has implications for human in vitro fertilization. We propose the zygotic checkpoint senses DNA lesions generated during paternal DNA demethylation and ensures reprogrammed loci are repaired before mitosis to prevent chromosome fragmentation, embryo loss, and infertility.
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, Ready Made Solution, 10 mg/mL in DMSO
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
NU7026, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
VE-821, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
KU-55933, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
AZD-7762 hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Caffeine, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-RAD21 Antibody, Upstate®, from mouse
Pricing and availability is not currently available.