Merck
  • Cortical development in the presenilin-1 null mutant mouse fails after splitting of the preplate and is not due to a failure of reelin-dependent signaling.
Cortical development in the presenilin-1 null mutant mouse fails after splitting of the preplate and is not due to a failure of reelin-dependent signaling.
Developmental dynamics : an official publication of the American Association of Anatomists (2008-08-30)
Rita De Gasperi, Miguel A Gama Sosa, Paul H Wen, Jingjun Li, Gissel M Perez, Tom Curran, Gregory A Elder
ABSTRACT
Cortical development is disrupted in presenilin-1 null mutant (Psen1-/-) mice. Prior studies have commented on similarities between Psen1-/- and reeler mice. Reelin induces phosphorylation of Dab1 and activates the phosphatidylinositol 3-kinase (PI3K)/Akt pathway. Psen1 is known to modulate PI3K/Akt signaling and both known reelin receptors (apoER2 and VLDLR) are substrates for Psen1 associated gamma-secretase activity. The purpose of this study was to determine whether reelin signaling is disrupted in Psen1-/- mice. We show that, while Dab1 is hypophosphorylated late in cortical development in Psen1-/- mice, it is normally phosphorylated at earlier ages and reelin signaling is intact in Psen1-/- primary neuronal cultures. gamma-secretase activity was also not required for reelin-induced phosphorylation of Dab1. Unlike reeler mice the preplate splits in Psen1-/- brain. Thus cortical development in Psen1-/- mice fails only after splitting of the preplate and is not due to an intrinsic failure of reelin signaling.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse