Merck
  • Selective Laminin-Directed Differentiation of Human Induced Pluripotent Stem Cells into Distinct Ocular Lineages.
Selective Laminin-Directed Differentiation of Human Induced Pluripotent Stem Cells into Distinct Ocular Lineages.
Cell reports (2018-11-08)
Shun Shibata, Ryuhei Hayashi, Toru Okubo, Yuji Kudo, Tomohiko Katayama, Yuki Ishikawa, Junko Toga, Emiko Yagi, Yoichi Honma, Andrew J Quantock, Kiyotoshi Sekiguchi, Kohji Nishida
ABSTRACT
The extracellular matrix plays a key role in stem cell maintenance, expansion, and differentiation. Laminin, a basement membrane protein, is a widely used substrate for cell culture including the growth of human induced pluripotent stem cells (hiPSCs). Here, we show that different isoforms of laminin lead to the selective differentiation of hiPSCs into different eye-like tissues. Specifically, the 211 isoform of the E8 fragment of laminin (LN211E8) promotes differentiation into neural crest cells via Wnt activation, whereas LN332E8 promotes differentiation into corneal epithelial cells. The immunohistochemical distributions of these laminin isoforms in the developing mouse eye mirrors the hiPSC type that was induced in vitro. Moreover, LN511E8 enables generation of dense hiPSC colonies due to actomyosin contraction, which in turn led to cell density-dependent YAP inactivation and subsequent retinal differentiation in colony centers. Thus, distinct laminin isoforms determine the fate of expanded hiPSCs into eye-like tissues.
Sigma-Aldrich
Anti-phospho-Vinculin (pTyr822) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.