Merck
  • Mutations in ARL2BP, a protein required for ciliary microtubule structure, cause syndromic male infertility in humans and mice.
Mutations in ARL2BP, a protein required for ciliary microtubule structure, cause syndromic male infertility in humans and mice.
PLoS genetics (2019-08-20)
Abigail R Moye, Nicola Bedoni, Jessica G Cunningham, Urikhan Sanzhaeva, Eric S Tucker, Peter Mathers, Virginie G Peter, Mathieu Quinodoz, Liliana P Paris, Luísa Coutinho-Santos, Pedro Camacho, Madeleine G Purcell, Abbie C Winkelmann, James A Foster, Elena N Pugacheva, Carlo Rivolta, Visvanathan Ramamurthy
ABSTRACT
Cilia are evolutionarily conserved hair-like structures with a wide spectrum of key biological roles, and their dysfunction has been linked to a growing class of genetic disorders, known collectively as ciliopathies. Many strides have been made towards deciphering the molecular causes for these diseases, which have in turn expanded the understanding of cilia and their functional roles. One recently-identified ciliary gene is ARL2BP, encoding the ADP-Ribosylation Factor Like 2 Binding Protein. In this study, we have identified multiple ciliopathy phenotypes associated with mutations in ARL2BP in human patients and in a mouse knockout model. Our research demonstrates that spermiogenesis is impaired, resulting in abnormally shaped heads, shortened and mis-assembled sperm tails, as well as in loss of axonemal doublets. Additional phenotypes in the mouse included enlarged ventricles of the brain and situs inversus. Mouse embryonic fibroblasts derived from knockout animals revealed delayed depolymerization of primary cilia. Our results suggest that ARL2BP is required for the structural maintenance of cilia as well as of the sperm flagellum, and that its deficiency leads to syndromic ciliopathy.
Sigma-Aldrich
Anti-SPEF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture