Merck
  • iNOS as a Driver of Inflammation and Apoptosis in Mouse Skeletal Muscle after Burn Injury: Possible Involvement of Sirt1 S-Nitrosylation-Mediated Acetylation of p65 NF-κB and p53.
iNOS as a Driver of Inflammation and Apoptosis in Mouse Skeletal Muscle after Burn Injury: Possible Involvement of Sirt1 S-Nitrosylation-Mediated Acetylation of p65 NF-κB and p53.
PloS one (2017-01-19)
Harumasa Nakazawa, Kyungho Chang, Shohei Shinozaki, Takashi Yasukawa, Kazuhiro Ishimaru, Shingo Yasuhara, Yong-Ming Yu, J A Jeevendra Martyn, Ronald G Tompkins, Kentaro Shimokado, Masao Kaneki
ABSTRACT
Inflammation and apoptosis develop in skeletal muscle after major trauma, including burn injury, and play a pivotal role in insulin resistance and muscle wasting. We and others have shown that inducible nitric oxide synthase (iNOS), a major mediator of inflammation, plays an important role in stress (e.g., burn)-induced insulin resistance. However, it remains to be determined how iNOS induces insulin resistance. Moreover, the interrelation between inflammatory response and apoptosis is poorly understood, although they often develop simultaneously. Nuclear factor (NF)-κB and p53 are key regulators of inflammation and apoptosis, respectively. Sirt1 inhibits p65 NF-κB and p53 by deacetylating these transcription factors. Recently, we have shown that iNOS induces S-nitrosylation of Sirt1, which inactivates Sirt1 and thereby increases acetylation and activity of p65 NF-κB and p53 in various cell types, including skeletal muscle cells. Here, we show that iNOS enhances burn-induced inflammatory response and apoptotic change in mouse skeletal muscle along with S-nitrosylation of Sirt1. Burn injury induced robust expression of iNOS in skeletal muscle and gene disruption of iNOS significantly inhibited burn-induced increases in inflammatory gene expression and apoptotic change. In parallel, burn increased Sirt1 S-nitrosylation and acetylation and DNA-binding capacity of p65 NF-κB and p53, all of which were reversed or ameliorated by iNOS deficiency. These results indicate that iNOS functions not only as a downstream effector but also as an upstream enhancer of burn-induced inflammatory response, at least in part, by Sirt1 S-nitrosylation-dependent activation (acetylation) of p65 NF-κB. Our data suggest that Sirt1 S-nitrosylation may play a role in iNOS-mediated enhanced inflammatory response and apoptotic change, which, in turn, contribute to muscle wasting and supposedly to insulin resistance after burn injury.
Sigma-Aldrich
Anti-Interferon-β Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Sirt1(Sir2) Antibody, Upstate®, from rabbit