Merck
  • Role of 5-ht7 receptors in the long-lasting hypotensive response induced by 5-hydroxytryptamine in the rat.
Role of 5-ht7 receptors in the long-lasting hypotensive response induced by 5-hydroxytryptamine in the rat.
British journal of pharmacology (1997-06-01)
J A Terrón
ABSTRACT
1. The receptor mediating the long-lasting hypotensive effect of intravenous (i.v.) 5-hydroxytryptamine (5-HT) in the rat was originally classified as 5-HT1-like. Since some pharmacological properties of this receptor are closely similar to those for the cloned 5-ht7 receptor, the present study investigated the effects of several 5-HT receptor agonists and antagonists showing high affinity for the cloned 5-ht7 receptor in pithed rats with artificially raised blood pressure. 2. I.v. bolus administration of 5-HT, 5-carboxamidotryptamine (5-CT), 5-methoxytryptamine, lisuride and sumatriptan to bilaterally vagotomized pithed rats pretreated with ketanserin (0.18 mumol kg-1, i.v.), the diastolic blood pressure of which had been raised by a continuous i.v. infusion of methoxamine (60-80 nmol kg-1 min-1), produced dose-dependent hypotensive responses; only 5-HT and 5-CT displayed similar maximum effects. In addition to mimicking the hypotensive action of 5-HT with a lower maximum effect, lisuride strongly antagonized the 5-CT-induced hypotensive responses thus suggesting a partial agonist effect. The rank order of hypotensive agonist potency was 5-CT > > 5-HT > or = 5-methoxytryptamine > or = lisuride > > sumatriptan. 3. In experiments with antagonists, i.v. treatment with metergoline (2.48 mumol kg-1), mesulergine (2.76 mumol kg-1), methysergide (2.13 mumol kg-1), lisuride (0.22 mumol kg-1), methiothepin (0.68 mumol kg-1), mianserin (10.6 mumol kg-1), or the atypical antipsychotic drugs, clozapine (11.0 mumol kg-1) or risperidone (78.0 nmol kg-1), produced significant rightward displacements of the dose-response curve for 5-CT in methoxamine-infused pithed animals pretreated with ketanserin (0.18 mumol kg-1, i.v.); lisuride, methiothepin and risperidone behaved as non-competitive antagonists as they elicited a significant reduction of the maximum effect to 5-CT. In contrast, blockade of 5-HT1, 5-HT3 and 5-HT4 receptors with i.v. propranolol (3.38 mumol kg-1), MDL-72222 (1.59 mumol kg-1) and GR125487 (1.91 mumol kg-1), respectively, did not alter 5-CT-induced hypotensive responses; ketanserin (0.18 mumol kg-1, i.v.) failed to modify the dose-response curve for 5-CT in saline-pretreated animals. Lastly, inhibition of the prostaglandin-forming cyclo-oxygenase and nitric oxide synthase with indomethacin (14 mumol kg-1, i.v.) and NG-nitro-L-arginine methyl ester (L-NAME, 120 mumol kg-1, i.v.), respectively, had no significant effects on 5-CT-induced hypotensive effects. 4. Taken together, the present pharmacological data suggest that the long-lasting vasodepressor action of 5-HT in the rat involves activation of receptors closely similar to the cloned 5-ht7 subtype. Since no evidence for an indirect mechanism could be obtained, these receptors may be primarily located in the vascular smooth muscle of the systemic resistance vessels. These findings represent further evidence favouring the functional role of the 5-ht7 receptor.