Merck
  • A Crucial Role for Ergosterol in Plasma Membrane Composition, Localisation, and Activity of Cdr1p and H+-ATPase in Candida albicans.
A Crucial Role for Ergosterol in Plasma Membrane Composition, Localisation, and Activity of Cdr1p and H+-ATPase in Candida albicans.
Microorganisms (2019-09-25)
Jakub Suchodolski, Jakub Muraszko, Przemysław Bernat, Anna Krasowska
ABSTRACT
Candida albicans is an opportunistic fungal pathogen of humans. Treatment of C. albicans infections relies on azoles, which target the lanosterol 14α-demethylase (Erg11p) encoded by the ERG11 gene. Our results show that targeted gene disruption of ERG11 can result in resistance to ergosterol-dependent drugs (azoles and amphotericin B), auxotrophy and aerobically viable erg11Δ/Δ cells. Abnormal sterol deposition and lack of ergosterol in the erg11Δ/Δ strain leads to reduced plasma membrane (PM) fluidity, as well as dysfunction of the vacuolar and mitochondrial membranes, resulting respectively in defects in vacuole fusion and a reduced intracellular ATP level. The altered PM structure of the erg11Δ/Δ strain contributes to delocalisation of H+-ATPase and the Cdr1 efflux pump from the PM to vacuoles and, resulting in a decrease in PM potential (Δψ) and increased sensitivity to ergosterol-independent xenobiotics. This new insight into intracellular processes under Erg11p inhibition may lead to a better understanding of the indirect effects of azoles on C. albicans cells and the development of new treatment strategies for resistant infections.
Product Description
Hamilton® MiniTrode lab pH electrode, glass, S7 connector
Sigma-Aldrich
Trifluoroacetamide, 97%