Merck
  • The phosphatidylinositol 3' kinase pathway is required for the survival signal of leukocyte tyrosine kinase.
The phosphatidylinositol 3' kinase pathway is required for the survival signal of leukocyte tyrosine kinase.
Oncogene (1997-06-26)
H Ueno, H Honda, T Nakamoto, T Yamagata, K Sasaki, K Miyagawa, K Mitani, Y Yazaki, H Hirai
ABSTRACT
Leukocyte tyrosine kinase (LTK) is a receptor tyrosine kinase which belongs to the insulin receptor superfamily and is mainly expressed in pre-B lymphocytes and neuronal tissues. Recently, we demonstrated that LTK utilizes Shc and IRS-1 as two major substrates and while both equally activate the Ras pathway, only IRS-1 suppresses apoptosis of hematopoietic cells, suggesting the existence of another unidentified signaling pathway downstream of IRS-1, which is relevant to the anti-apoptotic activity. In the present study, we found that wortmannin, a specific inhibitor of phosphatidylinositol 3' (PI3)-kinase, abolished the survival effects of LTK. Although c-Cbl is found to be phosphorylated by LTK and therefore is a second candidate linking LTK with the PI3-kinase pathway along with IRS-1, we found that the p85 subunit of PI3 kinase directly binds to tyrosine 753 of LTK, which is located within a YXXM motif, a consensus binding amino acid sequence for the SH2 domain of p85, but fails to bind to IRS-1 or c-Cbl. Ba/F3 cells which stably express the EGF receptor-LTK chimeric receptor carrying a mutation at tyrosine 753 fell into apoptotic death even in the presence of EGF, indicating that the PI3 kinase pathway is required for the survival effects of LTK.
Sigma-Aldrich
Phosphoinositides sodium salt from bovine brain
Pricing and availability is not currently available.