Merck
  • Interaction of nitrofurantoin with lipid langmuir monolayers as cellular membrane models distinguished with tensiometry and infrared spectroscopy.
Interaction of nitrofurantoin with lipid langmuir monolayers as cellular membrane models distinguished with tensiometry and infrared spectroscopy.
Colloids and surfaces. B, Biointerfaces (2020-01-24)
André C Machado, Luciano Caseli
ABSTRACT
Knowing how a drug interacts with cell membranes is important to understand and predict its effects at the molecular level. Therefore, we aimed to study the interaction of nitrofurantoin (NFT), a compound with potential antibiotic and antitumor properties, with lipidic biological interfaces using Langmuir monolayers. We employed the phospholipids 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and 1,2-dipalmitoyl-sn-glycero-3-phospho-l-serine (DPPS), which were spread on the surface of water to form Langmuir films, to investigate the membrane-drug interactions. The interaction of the drug with the lipid monolayers was evaluated by using surface pressure-area isotherms, surface pressure-time kinetic curves, Brewster angle microscopy (BAM), and polarization-modulated infrared reflection-absorption spectroscopy (PM-IRRAS). Nitrofurantoin shifted the isotherms to lower DPPC molecular areas, indicating monolayer condensation, and to higher DPPS molecular areas, indicating monolayer expansion. Meanwhile, BAM images showed the appearance of interfacial aggregates for DPPS, but not for DPPC, in the presence of NFT. PM-IRRAS spectra showed that bands related to methylene stretches changed their relative intensities and maximum position related to their asymmetric and symmetric modes for both lipids. This suggested an alteration of the monolayer packing degree and the conformational order of the lipid alkyl chains, which were related to an increase in configurational order for DPPS, but disorder for DPPC. In conclusion, NFT caused distinctive changes in the thermodynamic, morphological, and structural properties of DPPC and DPPS monolayers, which may be associated with its bioactivity in cellular membranes and other lipidic interfaces of pharmaceutical interest.
Product Description
Avanti
16:0 PS, Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand