Merck
  • LRRTMs Organize Synapses through Differential Engagement of Neurexin and PTPσ.
LRRTMs Organize Synapses through Differential Engagement of Neurexin and PTPσ.
Neuron (2020-01-30)
Reiko T Roppongi, Shreya H Dhume, Nirmala Padmanabhan, Prabhisha Silwal, Nazmeena Zahra, Benyamin Karimi, Claire Bomkamp, Chetan S Patil, Kevin Champagne-Jorgensen, Rebecca E Twilley, Peng Zhang, Michael F Jackson, Tabrez J Siddiqui
ABSTRACT
Presynaptic neurexins (Nrxs) and type IIa receptor-type protein tyrosine phosphatases (RPTPs) organize synapses through a network of postsynaptic ligands. We show that leucine-rich-repeat transmembrane neuronal proteins (LRRTMs) differentially engage the protein domains of Nrx but require its heparan sulfate (HS) modification to induce presynaptic differentiation. Binding to the HS of Nrx is sufficient for LRRTM3 and LRRTM4 to induce synaptogenesis. We identify mammalian Nrx1γ as a potent synapse organizer and reveal LRRTM4 as its postsynaptic ligand. Mice expressing a mutant form of LRRTM4 that cannot bind to HS show structural and functional deficits at dentate gyrus excitatory synapses. Through the HS of Nrx, LRRTMs also recruit PTPσ to induce presynaptic differentiation but function to varying degrees in its absence. PTPσ forms a robust complex with Nrx, revealing an unexpected interaction between the two presynaptic hubs. These findings underscore the complex interplay of synapse organizers in specifying the molecular logic of a neural circuit.
Sigma-Aldrich
7-Amino-4-methyl-3-coumarinylacetic acid, BioReagent, suitable for fluorescence, ~90% (HPLC)
Sigma-Aldrich
Heparinase III from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% (w/w) bovine serum albumin, ≥30 units/mg protein (enzyme + BSA)
Sigma-Aldrich
Heparinase I from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% bovine serum albumin, ≥200 units/mg protein (enzyme + BSA)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Tau-1 Antibody, clone PC1C6, clone PC1C6, Chemicon®, from mouse
Sigma-Aldrich
Anti-pan Neurexin-1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Heparinase II from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% bovine serum albumin, lyophilized powder, ≥100 units/mg protein (enzyme + BSA)
Avanti
n-dodecyl-β-D-maltoside (DDM), Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Synapsin I Antibody, Chemicon®, from rabbit