Merck
  • Liposomal Lapatinib in Combination with Low-Dose Photodynamic Therapy for the Treatment of Glioma.
Liposomal Lapatinib in Combination with Low-Dose Photodynamic Therapy for the Treatment of Glioma.
Journal of clinical medicine (2019-12-19)
Carl Fisher, Girgis Obaid, Carolyn Niu, Warren Foltz, Alyssa Goldstein, Tayyaba Hasan, Lothar Lilge
ABSTRACT
Malignant gliomas are highly invasive and extremely difficult to treat tumours with poor prognosis and outcomes. Photodynamic therapy (PDT), mediated by Gleolan®, has been studied previously with partial success in treating these tumours and extending lifetime. We aim to determine whether combining PDT using ALA-protoporphyrin IX (PpIX) with a liposomal formulation of the clinical epidermal growth factor receptor (EGFR) inhibitor, lapatinib, would increase the anti-tumour PDT efficacy. Lapatinib was given in vitro and in vivo 24 h prior to PDT and for 3-5 days following PDT to elicit whether the combination provided any benefits to PDT therapy. Live-cell imaging, in vitro PDT, and in vivo studies were performed to elucidate the effect lapatinib had on PDT for a variety of glioma cell lines and as well as GSC-30 neurospheres in vivo. PDT combined with lapatinib led to a significant increase in PpIX accumulation, and reductions in the LD50 of PpIX mediated PDT in two EGFR-driven cell lines, U87 and U87vIII, tested (p < 0.05). PDT + lapatinib elicited stronger MRI-quantified glioma responses following PDT for two human glioma-derived tumours (U87 and GSC-30) in vivo (p < 0.05). Furthermore, PDT leads to enhanced survival in rats following treatment with lapatinib compared to lapatinib alone and PDT alone (p < 0.05). As lapatinib is approved for other oncological indications, a realization of its potential combination with PDT and in fluorescence-guided resection could be readily tested clinically. Furthermore, as its use would only be in acute settings, long-term resistance should not pose an issue as compared to its use as monotherapy.
Sigma-Aldrich
Lapatinib
Avanti
Cholesterol (ovine), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine, ≥99%
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand