Merck
  • Ampakines Attenuate Staurosporine-induced Cell Death in Primary Cortical Neurons: Implications in the 'Chemo-Brain' Phenomenon.
Ampakines Attenuate Staurosporine-induced Cell Death in Primary Cortical Neurons: Implications in the 'Chemo-Brain' Phenomenon.
Anticancer research (2018-06-01)
Daniel P Radin, Gary A Rogers, Kimberley E Hewitt, Richard Purcell, Arnold Lippa
ABSTRACT
Mounting evidence suggests that trophic cell signaling can be mediated by alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) activation. It has been demonstrated that exogenous application of brain-derived neurotrophic factor (BDNF) is highly neuroprotective in vitro against neurotoxic insults such as standard chemotherapies. Because positive allosteric modulation of AMPARs with ampakines can increase both BDNF mRNA and protein in vitro and in vivo, we examined whether application of the ampakines CX614 and CX729 promoted neuroprotection against staurosporine-induced cell death in rat primary cortical neurons using propidium iodide to stain for dead cells. A transient 2-h pretreatment with CX614 or CX729 performed 24 h prior to staurosporine produced significant, time-dependent neuroprotection that was resistant to the AMPAR antagonists NBQX or GYKI 52466, suggesting that this effect may be independent of ion flow. Furthermore, the pretreatment time requirements of CX729 matched the time course for increased BDNF expression previously reported to occur in hippocampal slices, suggesting that increased neurotrophin expression might be associated with the neuroprotective effects conferred by ampakines. Our data demonstrate that ampakines may be able to perturb neuronal toxicity and peripheral neuropathy of front-line chemotherapies.
Sigma-Aldrich
CX614, ≥98% (HPLC)