Merck
  • Membrane fission is promoted by insertion of amphipathic helices and is restricted by crescent BAR domains.
Membrane fission is promoted by insertion of amphipathic helices and is restricted by crescent BAR domains.
Cell (2012-04-03)
Emmanuel Boucrot, Adi Pick, Gamze Çamdere, Nicole Liska, Emma Evergren, Harvey T McMahon, Michael M Kozlov
ABSTRACT
Shallow hydrophobic insertions and crescent-shaped BAR scaffolds promote membrane curvature. Here, we investigate membrane fission by shallow hydrophobic insertions quantitatively and mechanistically. We provide evidence that membrane insertion of the ENTH domain of epsin leads to liposome vesiculation, and that epsin is required for clathrin-coated vesicle budding in cells. We also show that BAR-domain scaffolds from endophilin, amphiphysin, GRAF, and β2-centaurin limit membrane fission driven by hydrophobic insertions. A quantitative assay for vesiculation reveals an antagonistic relationship between amphipathic helices and scaffolds of N-BAR domains in fission. The extent of vesiculation by these proteins and vesicle size depend on the number and length of amphipathic helices per BAR domain, in accord with theoretical considerations. This fission mechanism gives a new framework for understanding membrane scission in the absence of mechanoenzymes such as dynamin and suggests how Arf and Sar proteins work in vesicle scission.
Product Description
Avanti
Brain Extract Total, Avanti Research - A Croda Brand
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain Extract Total, Avanti Research - A Croda Brand