Merck
  • Lysophosphatidylcholine-induced mitochondrial fission contributes to collagen production in human cardiac fibroblasts.
Lysophosphatidylcholine-induced mitochondrial fission contributes to collagen production in human cardiac fibroblasts.
Journal of lipid research (2019-08-01)
Hui-Ching Tseng, Chih-Chung Lin, Li-Der Hsiao, Chuen-Mao Yang
ABSTRACT
Lysophosphatidylcholine (LPC) may accumulate in the heart to cause fibrotic events, which is mediated through fibroblast activation and collagen accumulation. Here, we evaluated the mechanisms underlying LPC-mediated collagen induction via mitochondrial events in human cardiac fibroblasts (HCFs), coupling application of the pharmacologic cyclooxygenase-2 (COX-2) inhibitor, celecoxib, and genetic mutations in FOXO1 on the fibrosis pathway. In HCFs, LPC caused prostaglandin E2 (PGE2)/PGE2 receptor 4 (EP4)-dependent collagen induction via activation of transcriptional activity of forkhead box protein O1 (FoxO1) on COX-2 gene expression. These responses were mediated through LPC-induced generation of mitochondrial reactive oxygen species (mitoROS), as confirmed by ex vivo studies, which indicated that LPC increased COX-2 expression and oxidative stress. LPC-induced mitoROS mediated the activation of protein kinase C (PKC)α, which interacted with and phosphorylated dynamin-related protein 1 (Drp1) at Ser616, thereby increasing Drp1-mediated mitochondrial fission and mitochondrial depolarization. Furthermore, inhibition of PKCα and Drp1 reduced FoxO1-mediated phosphorylation at Ser256 and nuclear accumulation, which suppressed COX-2/PGE2 expression and collagen production. Moreover, pretreatment with celecoxib or COX-2 siRNA suppressed WT FoxO1; mutated Ser256-to-Asp256 FoxO1-enhanced collagen induction, which was reversed by addition of PGE2 Our results demonstrate that LPC-induced generation of mitoROS regulates PKCα-mediated Drp1-dependent mitochondrial fission and COX-2 expression via a PKCα/Drp1/FoxO1 cascade, leading to PGE2/EP4-mediated collagen induction. These findings provide new insights about the role of LPC in the pathway of fibrotic injury in HCFs.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid
Sigma-Aldrich
L-α-Lysophosphatidylcholine from Glycine max (soybean), ≥99%, lyophilized powder
Sigma-Aldrich
Mitochondria Staining Kit, 1 kit sufficient for 40 tests (of 5 mL cell suspensions), 1 kit sufficient for 200 tests (of 1 mL cell suspensions)