Merck
  • 4F-MDMB-BINACA: A New Synthetic Cannabinoid Widely Implicated in Forensic Casework.
4F-MDMB-BINACA: A New Synthetic Cannabinoid Widely Implicated in Forensic Casework.
Journal of forensic sciences (2019-07-02)
Alex J Krotulski, Amanda L A Mohr, Sherri L Kacinko, Melissa F Fogarty, Sarah A Shuda, Francis X Diamond, William A Kinney, M J Menendez, Barry K Logan
ABSTRACT
This is the first report regarding the characterization of the new synthetic cannabinoid 4F-MDMB-BINACA. 4F-MDMB-BINACA was first analytically confirmed in seized drug material using gas chromatography-mass spectrometry (GC-MS), liquid chromatography-quadrupole time-of-flight mass spectrometry (LC-QTOF), and nuclear magnetic resonance (NMR) spectroscopy. Subsequent to this characterization, 4F-MDMB-BINACA was detected in biological specimens collected as part of forensically relevant casework, including medicolegal death investigations and drug impaired driving investigations, from a variety of regions in the United States. Further analysis of biological specimens resulted in the identification of the metabolites 4F-MDMB-BINACA 3,3-dimethylbutanoic acid and 4-OH-MDMB-BINACA. 4F-MDMB-BINACA is appearing with increasing frequency as a contributory factor in deaths, creating morbidity and mortality risks for drug users. Laboratories must be aware of its presence and impact, incorporating 4F-MDMB-BINACA into workflows for detection and confirmation.
Sigma-Aldrich
3,3-Dimethylbutyric acid, 98%
Pricing and availability is not currently available.