Merck
  • Quality and Consumer Acceptance of Meat from Rabbits Fed Diets in Which Soybean Oil is Replaced with Black Soldier Fly and Yellow Mealworm Fats.
Quality and Consumer Acceptance of Meat from Rabbits Fed Diets in Which Soybean Oil is Replaced with Black Soldier Fly and Yellow Mealworm Fats.
Animals : an open access journal from MDPI (2019-09-01)
Laura Gasco, Sihem Dabbou, Francesco Gai, Alberto Brugiapaglia, Achille Schiavone, Marco Birolo, Gerolamo Xiccato, Angela Trocino
ABSTRACT
This trial investigated the effect of the dietary inclusion of Hermetia illucens (H) and Tenebrio molitor (T) fats as alternative lipid sources for growing rabbits, and assessed the carcass characteristics; proximate composition; lipid peroxidation, and fatty acid profile of the meat, as well as consumer acceptance. At weaning, 200 crossbred rabbits (1051 ± 138 g initial body weight) were allotted to five isolipidic (4% dry matter (DM)) dietary treatments: a control diet (C) containing 1.5% of soybean oil, and four experimental diets in which soybean oil was partially (50%) or totally (100%) substituted by H (H50 and H100) or T (T50 and T100) fats. The carcass characteristics, the meat quality traits, and the consumer acceptance of the cooked meat were not affected. The fat content of Longissimus thoracis et lumborum muscle of the rabbits was 1.1% on average. In the case of rabbit fed the H diets (average of diets H50 and H100), the same muscles revealed a higher saturated fatty acid proportion (47.1% vs. 39.7% and 40.8%, respectively) and a lower polyunsaturated fatty acid proportion than the rabbits fed the C and T diets (average of diets T50 and T100) (26.5% vs. 31.7% and 29.7%) (p < 0.001). The meat of the rabbits fed the diets containing insect fat (average for H and T diets) was less susceptible to oxidation (0.24 vs. 0.39 mg malondialdehyde/kg meat in the C group; p < 0.01).
Sigma-Aldrich
1,1,3,3-Tetramethoxypropane, 99%
Pricing and availability is not currently available.