Merck
  • Muscarinic acetylcholine receptor regulates self-renewal of early erythroid progenitors.
Muscarinic acetylcholine receptor regulates self-renewal of early erythroid progenitors.
Science translational medicine (2019-09-27)
Gaurang Trivedi, Daichi Inoue, Cynthia Chen, Lillian Bitner, Young Rock Chung, Justin Taylor, Mithat Gönen, Jürgen Wess, Omar Abdel-Wahab, Lingbo Zhang
ABSTRACT
Adult stem and progenitor cells are uniquely capable of self-renewal, and targeting this process represents a potential therapeutic opportunity. The early erythroid progenitor, burst-forming unit erythroid (BFU-E), has substantial self-renewal potential and serves as a key cell type for the treatment of anemias. However, our understanding of mechanisms underlying BFU-E self-renewal is extremely limited. Here, we found that the muscarinic acetylcholine receptor, cholinergic receptor, muscarinic 4 (CHRM4), pathway regulates BFU-E self-renewal and that pharmacological inhibition of CHRM4 corrects anemias of myelodysplastic syndrome (MDS), aging, and hemolysis. Genetic down-regulation of CHRM4 or pharmacologic inhibition of CHRM4 using the selective antagonist PD102807 promoted BFU-E self-renewal, whereas deletion of Chrm4 increased erythroid cell production under stress conditions in vivo. Moreover, muscarinic acetylcholine receptor antagonists corrected anemias in mouse models of MDS, aging, and hemolysis in vivo, extending the survival of mice with MDS relative to that of controls. The effects of muscarinic receptor antagonism on promoting expansion of BFU-Es were mediated by cyclic AMP induction of the transcription factor CREB, whose targets up-regulated key regulators of BFU-E self-renewal. On the basis of these data, we propose a model of hematopoietic progenitor self-renewal through a cholinergic-mediated "hematopoietic reflex" and identify muscarinic acetylcholine receptor antagonists as potential therapies for anemias associated with MDS, aging, and hemolysis.
Sigma-Aldrich
KT 5720, ≥98% (HPLC), powder
Supelco
Orphenadrine citrate salt, analytical standard
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Phenylhydrazine, 97%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human KIT
Sigma-Aldrich
MISSION® esiRNA, targeting human CREB1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Zfp36l2
Sigma-Aldrich
ChIPAb+ CREB - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, clone NL904, from rabbit
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human GATA2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chrm4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gata2
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O