Merck
  • Determination of serum imatinib and its' metabolite in patients chronic myeloid leukemia.
Determination of serum imatinib and its' metabolite in patients chronic myeloid leukemia.
Clinica chimica acta; international journal of clinical chemistry (2019-07-28)
Duygu Eryavuz Onmaz, Sedat Abusoglu, Ali Unlu, Abdulkadir Basturk, Mehmet Dagli, Metin Bagci, Oguzhan Tok, Gulsum Abusoglu
ABSTRACT
Imatinib has favorable pharmacokinetic properties, but primary and secondary resistance mechanisms may cause a decrease in clinical response over time. There is a positive correlation between serum imatinib concentrations and treatment response. Our aim was to develop a method for the measurement of imatinib and its' active metabolite N-desmethyl imatinib. Serum imatinib and N-desmethyl imatinib levels were determined by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and validation studies were carried out according to CLSI (The Clinical & Laboratory Standards Institute) protocols. Serum samples were collected from 40 patients with chronic myeloid leukemia (CML) and analyzed with LC-MS/MS and ultra high-performance liquid chromatography (UHPLC) methods. The linearity range and correlation coefficient were 12.2-12,500 ng/mL and 0.9987 for LC-MS/MS method, respectively. Limit of quantitation was determined as 24.4 ng/mL. The retention times of imatinib and N-desmethyl imatinib were 1.66 and 1.60 min, respectively. There was no statistically significant difference between the results of both methods. This LC-MS/MS method is cost-effective and has adavantages such as using low serum volumes, requiring simple pretreatment steps (only protein precipitation) and reduced turnaround times for analysis.
Product Description
Supelco
Acetonitrile, for UHPLC-MS LiChrosolv®
Sigma-Aldrich
Acetonitrile, for UHPLC, suitable for mass spectrometry (MS)
Sigma-Aldrich
Imatinib mesylate