Merck
  • Connexin37-Dependent Mechanisms Selectively Contribute to Modulate Angiotensin II -Mediated Hypertension.
Connexin37-Dependent Mechanisms Selectively Contribute to Modulate Angiotensin II -Mediated Hypertension.
Journal of the American Heart Association (2019-04-05)
Loïc Le Gal, Maxime Pellegrin, Tania Santoro, Lucia Mazzolai, Armin Kurtz, Paolo Meda, Charlotte Wagner, Jacques-Antoine Haefliger
ABSTRACT
Background Gap junction channels made of Connexin37 (Cx37) are expressed by aortic endothelial and smooth muscle cells of hypertensive mice, as well as by the renin-secreting cells of kidneys. Methods and Results To decipher whether Cx37 has any role in hypertension, angiotensin II (Ang II ) was infused in normotensive wild-type and Cx37-deficient mice (Cx37-/-). After 2 to 4 weeks, the resulting increase in blood pressure was lower in Cx37-/- than in wild-type mice, suggesting an alteration in the Ang II response. To investigate this possibility, mice were submitted to a 2-kidney, 1-clip procedure, a renin-dependent model of hypertension. Two weeks after this clipping, Cx37-/- mice were less hypertensive than wild-type mice and, 2 weeks later, their blood pressure had returned to control values, in spite of abnormally high plasma renin levels. In contrast, Cx37-/- and wild-type mice that received N-nitro-l-arginine-methyl-ester, a renin-independent model of hypertension, featured a similar and sustained increase in blood pressure. The data indicate that loss of Cx37 selectively altered the Ang II -dependent pathways. Consistent with this conclusion, aortas of Cx37-/- mice featured an increased basal expression of the Ang II type 2 receptors ( AT 2R), and increased transcripts levels of downstream signaling proteins, such as Cnksr1 and Ptpn6 ( SHP -1). Accordingly, the response of Cx37-/- mice aortas to an ex vivo Ang II exposure was altered, since phosphorylation levels of several proteins of the Ang II pathway ( MLC 2, ERK , and AKT ) remained unchanged. Conclusions These findings provide evidence that Cx37 selectively influences Ang II signaling, mostly via a modulation of the expression of the Ang II type 2 receptor.
Sigma-Aldrich
Anti-Connexin 45 Antibody, near CT, cytoplasmic, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-Connexin 40 Antibody, Chemicon®, from rabbit