Merck
  • Ligand Effect in Alkali-Metal-Catalyzed Transfer Hydrogenation of Ketones.
Ligand Effect in Alkali-Metal-Catalyzed Transfer Hydrogenation of Ketones.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-07-19)
Iryna D Alshakova, Hayden C Foy, Travis Dudding, Georgii I Nikonov
ABSTRACT
This work unveils the reactivity patterns, as well as ligand and additive effect on alkali-metal-base-catalyzed transfer hydrogenation of ketones. Crucially to this reactivity is the presence of a Lewis acid (alkali cation), as opposed to a simple base effect. With aryl ketones, the observed reactivity order is Na+ >Li+ >K+ , whereas for aliphatic substrates it follows the expected Lewis acidity, Li+ >Na+ >K+ . Importantly, the reactivity pattern can be drastically changed by adding ligands and additives. Kinetic, labelling, and competition experiments as well as DFT calculations suggested that the reaction proceeds through a concerted direct hydride-transfer mechanism, originally suggested by Woodward. The lithium cation was found to be intrinsically more active than heavier congeners, but in the case of aryl ketones a decrease in reaction rate was observed at ≈40 % conversion with lithium cations. Noncovalent-interaction analysis revealed that this deceleration effect originated from specific noncovalent interactions between the aryl moiety of 1-phenylethanol and the carbonyl group of acetophenone, which stabilize the product in the coordination sphere of lithium and thus poison the catalyst. The ligand/additive effect is a complicated phenomenon that includes a combination of several factors, such as the decrease of activation energy by ligation (confirmed by distortion/interaction calculations of N,N,N',N'-tetramethylethylenediamine, TMEDA) and the change in relative stabilization of reagents and substrates in the solution and the coordination sphere of the metal. Finally, we observed that lithium-base-catalyzed transfer hydrogenation can be further facilitated by the addition of an inexpensive and benign reagent, LiCl, which likely operates by re-initiating the reaction on a new lithium center.
Sigma-Aldrich
2,6-Bis[(4R)-(+)-isopropyl-2-oxazolin-2-yl]pyridine, 99%
Pricing and availability is not currently available.