Merck
  • Polysaccharide enhanced NK cell cytotoxicity against pancreatic cancer via TLR4/MAPKs/NF-κB pathway in vitro/vivo.
Polysaccharide enhanced NK cell cytotoxicity against pancreatic cancer via TLR4/MAPKs/NF-κB pathway in vitro/vivo.
Carbohydrate polymers (2019-09-16)
Xin Xie, Lingman Ma, Yiran Zhou, Wen Shen, Duiyue Xu, Jie Dou, Baiyong Shen, Changlin Zhou
ABSTRACT
A polysaccharide isolated from Strongylocentrotus nudus eggs (SEP) reportedly displays immune activity in vivo. Here, its effect and underlying mechanism in the treatment of pancreatic cancer were investigated. SEP obviously inhibited pancreatic cancer growth by activating NK cells in vitro/vivo via TLR4/MAPKs/NF-κB signaling pathway, The tumor inhibitory rate achieved to 44.5% and 50.8% at a dose of 40 mg/kg in Bxpc-3 and SW1990 nude mice, respectively. Moreover, SEP obviously augmented the Gemcitabine (GEM) antitumor effect by upregulating NKG2D, which improved the sensitivity of NK cells targeting to its ligand MICA; meanwhile, the antitumor inhibitory rate was 68.6% in BxPC-3 tumor-bearing mice. Moreover, SEP reversed GEM-induced apoptosis and atrophy in both spleen and bone marrow via suppressing ROS secretion in vivo. These results suggested that pancreatic cancer was effectively inhibited by SEP-enhanced NK cytotoxicity mediated primarily through TLR4/MAPKs/NF-κB signaling pathway, representing a potential immunotherapy candidate for the treatment of pancreatic cancer.
Sigma-Aldrich
1-Cyclohexene-1-carboxylic acid, 97%
Pricing and availability is not currently available.