Merck
  • Isolation and Identification of Lactic Acid Bacteria Probiotic Culture Candidates for the Treatment of Salmonellaenterica Serovar Enteritidis in Neonatal Turkey Poults.
Isolation and Identification of Lactic Acid Bacteria Probiotic Culture Candidates for the Treatment of Salmonellaenterica Serovar Enteritidis in Neonatal Turkey Poults.
Animals : an open access journal from MDPI (2019-09-20)
Margarita A Arreguin-Nava, Daniel Hernández-Patlán, Bruno Solis-Cruz, Juan D Latorre, Xochitl Hernandez-Velasco, Guillermo Tellez, Saeed El-Ashram, Billy M Hargis, Guillermo Tellez-Isaias
ABSTRACT
The effect of Lactobacillus spp.-based probiotic candidates on Salmonella enterica serovar Enteritidis (SE) colonization was evaluated in two separate experiments. In each experiment, sixty-one day-of-hatch female turkey poults were obtained from a local hatchery. In both experiments, poults were challenged via oral gavage with 104 cfu/poult of SE and randomly allocated to one of two groups (n = 30 poults): (1) the positive control group and (2) the probiotic treated group. Heated brooder batteries were used for housing each group separately and poults were allowed ad libitum access to water and unmedicated turkey starter feed. 1 h following the SE challenge, poults were treated with 106 cfu/poult of probiotic culture via oral gavage or phosphate-buffered saline (PBS)to control groups. A total of 24 h post-treatment, poults were euthanized and the ceca and cecal tonsils from twenty poults were collected aseptically for SE recovery. In both trials, a significant reduction in the incidence and log10 cfu/g of SE were observed in poults treated with the probiotic when compared with control poults (p ≤ 0.05). The results of the present study suggest that the administration of this lactic acid-producing bacteria (LAB)-based probiotic 1 h after an SE challenge can be useful in reducing the cecal colonization of this pathogen in neonatal poults.
Product Description
Millipore
Brilliant green agar, modified, suitable for microbiology, NutriSelect® Basic
Pricing and availability is not currently available.
Sigma-Aldrich
Nalidixic acid sodium salt, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)
Pricing and availability is not currently available.