- Short-term effects of Vertical sleeve gastrectomy and Roux-en-Y gastric bypass on glucose homeostasis.
Short-term effects of Vertical sleeve gastrectomy and Roux-en-Y gastric bypass on glucose homeostasis.
The objective of this study was to compare the biochemical changes related to glucose tolerance and lipid metabolism in non-diabetic patients shortly after vertical sleeve gastrectomy (SG) or Roux-en-Y gastric bypass (RYGB). Non-diabetic women and men with morbid obesity were studied the day before and six days after SG (N = 15) or RYGB (N = 16). Patients completed an oral glucose tolerance test (OGTT; 75 g glucose) at both visits. SG and RYGB similarly improved fasting glucose homeostasis six days after surgery, with reduced glucose and insulin concentrations. The OGTT revealed differences between the two surgery groups that were not evident from the fasting serum concentrations. Postprandial (120 min) glucose and insulin concentrations were lower after RYGB but not after SG, whereas concentrations of glucagon-like peptide-1, peptide YY, glucagon and non-esterified fatty acids were elevated after both SG and RYGB. Fasting triacylglycerol concentration did not change after surgery, but concentrations of high density lipoprotein and low density lipoprotein cholesterols were reduced in both surgery groups, with no differences between the groups. To conclude, RYGB induced a more pronounced improvement in postprandial glucose homeostasis relative to SG, possibly due to improved insulin sensitivity rather than augmented insulin concentration.